Learn More
Invitrogen™ CBR1 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA518070
Product Code. 11316043
  • € 578.00 / 100µg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CBR1
Polyclonal
Unconjugated
CBR1
15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1
Goat
Ammonium sulfate precipitation
RUO
873
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P16152
CBR1
Synthetic peptide sequence (HGQFVSEKRVEQW) corresponding to the C-terminus amino acids of CBR1 (aa 265-277).