missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR3 Antibody (OTI1G6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70343PE
This item is not returnable.
View return policy
Description
CBR3 Monoclonal antibody specifically detects CBR3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CBR3 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1G6 | |
| Western Blot, Immunohistochemistry | |
| carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
| Full length human recombinant protein of human CBR3(NP_001227) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 874 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction