missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBS Recombinant Rabbit Monoclonal Antibody (1R2E9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA535140

Product Code. 17155803

  • € 488.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EDLTEKKPWW WHLRVQELGL SAPLTVLPTI TCGHTIEILR EKGFDQAPVV DEAGVILGMV TLGNMLSSLL AGKVQPSDQV GKVIYKQFKQ IRLTDTLGRL SHILEMDHFA LVVHEQIQYH STGKSSQRQM VFGVVTAIDL LNFVAAQERD QK.

Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CBS
Recombinant Monoclonal
0.63 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P32232, P35520, Q91WT9
CBS
Recombinant fusion protein containing a sequence corresponding to amino acids 400-551 of human CBS (P35520).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1R2E9
Unconjugated
CBS
AI047524; AI303044; Beta-thionase; Cbs; CBSL; cystathionine beta synthase; cystathionine beta-synthase; cystathionine beta-synthase; LOW QUALITY PROTEIN: cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; hemoprotein H-450; HIP4; LOC100057612; methylcysteine synthase; serine sulfhydrase
Rabbit
Affinity Chromatography
RUO
12411, 24250, 875
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.