missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBS Polyclonal Antibody

Product Code. 16344195 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16344195 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16344195 Supplier Invitrogen™ Supplier No. PA594923

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Human CBS shares 83% amino acid (aa) sequence identity with both mouse and rat CBS. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human K562 whole cell, rat liver tissue, rat pancrease tissue, mouse liver tissue, mouse pancrease tissue. IHC: human liver cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene CBS
Gene Accession No. P32232, P35520, Q91WT9
Gene Alias AI047524; AI303044; Beta-thionase; Cbs; CBSL; cystathionine beta synthase; cystathionine beta-synthase; cystathionine beta-synthase; LOW QUALITY PROTEIN: cystathionine beta-synthase; Cystathionine beta-synthase-like protein; cystathionine-beta-synthase; hemoprotein H-450; HIP4; LOC100057612; methylcysteine synthase; serine sulfhydrase
Gene Symbols CBS
Host Species Rabbit
Immunogen E.coli-derived human CBS recombinant protein (Position: A331-K551).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12411, 24250, 875
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.