missing translation for 'onlineSavingsMsg'
Learn More

CBS Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA598312

Additional Details : Weight : 0.01000kg

Product Code. 16501142

  • € 424.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Only known pyridoxal phosphate-dependent enzyme that contains heme. Important regulator of hydrogen sulfide, especially in the brain, utilizing cysteine instead of serine to catalyze the formation of hydrogen sulfide. Hydrogen sulfide is a gastratransmitter with signaling and cytoprotective effects such as acting as a neuromodulator in the brain to protect neurons against hypoxic injury.
Specifications

Specifications

CBS
Polyclonal
Unconjugated
CBS
AI047524, AI303044, Beta-thionase, Cbs, CBSL, cystathionine beta synthase, cystathionine beta-synthase, cystathionine beta-synthase, LOW QUALITY PROTEIN: cystathionine beta-synthase, Cystathionine beta-synthase-like protein, cystathionine-beta-synthase, hemoprotein H-450, HIP4, LOC100057612, methylcysteine synthase, serine sulfhydrase
Rabbit
Protein G
RUO
12411, 875
-20°C or -80°C if preferred
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot
3.75 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4
P35520, Q91WT9
CBS
Recombinant Human Cystathionine beta-synthase protein (1-413AA)
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers