Learn More
CBWD1 Monoclonal Antibody (OTI1B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501620
Encompass_Preferred
 View more versions of this product
Product Code. 17564731
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
CBWD1 is a protein coding gene.
Specifications
Specifications
CBWD1
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BRT8
CBWD1
Full length human recombit protein of human CBWD1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1B3
Unconjugated
CBWD1
COBP
Mouse
Affinity Chromatography
RUO
55871
-20° C, Avoid Freeze/Thaw Cycles