missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBWD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570998
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
CBWD3 is a protein coding gene.
Specifications
| CBWD3 | |
| Polyclonal | |
| Unconjugated | |
| CBWD3 | |
| bA561O23.1; CBWD3; Cobalamin synthase W domain-containing protein 3; cobalamin synthetase W domain-containing protein 3; COBW domain containing 3; COBW domain-containing protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 445571 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5JTY5 | |
| CBWD3 | |
| synthetic peptide directed towards the C-terminal region of human CBWD3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction