missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx2 Antibody (RAB-C238), Janelia Fluor™ 646 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20135JF646
This item is not returnable.
View return policy
Description
Cbx2 Monoclonal antibody specifically detects Cbx2 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| Cbx2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX2 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 84733 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunofluorescence | |
| RAB-C238 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction