missing translation for 'onlineSavingsMsg'
Learn More

CBX2 Monoclonal Antibody (4C11B10)
GREENER_CHOICE

Product Code. 17460783 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17460783 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17460783 Supplier Thermo Scientific Supplier No. MA538465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CBX2 Monoclonal Antibody (4C11B10)

CBX2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4C11B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CBX2
Gene Accession No. P30658, Q14781
Gene Alias Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5
Gene Symbols CBX2
Host Species Mouse
Immunogen Purified recombinant fragment of human CBX2 (aa: 402-525) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12416, 84733
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.