missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBX2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16600195 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16600195 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16600195 Supplier Invitrogen™ Supplier No. PA5103739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CBX2.

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CBX2
Gene Accession No. P30658, Q14781
Gene Alias Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5
Gene Symbols CBX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CBX2(Accession Q14781), corresponding to amino acid residues H258-I308.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12416, 84733
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.