missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530996
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
Specifications
| CBX2 | |
| Polyclonal | |
| Unconjugated | |
| CBX2 | |
| Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12416, 84733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.9 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P30658, Q14781 | |
| CBX2 | |
| Recombinant protein encompassing a sequence within the center region of human CBX2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction