missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBX2 Recombinant Rabbit Monoclonal Antibody (24A2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550021
This item is not returnable.
View return policy
Description
CBX2 Recombinant Monoclonal Antibody for Western Blot, ELISA
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
Specifications
| CBX2 | |
| Recombinant Monoclonal | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14781 | |
| CBX2 | |
| A synthesized peptide derived from human CBX2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 24A2 | |
| Unconjugated | |
| CBX2 | |
| Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 303730, 84733 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction