missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104496
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CBX6.
Polycomb group (PcG) proteins form multiprotein complexes that regulate expression patterns of developmental and cell proliferation genes. CBX6 (chromobox protein homolog 6) is a 412 amino acid protein that is a component of the chromatin-associated PcG complex. Through interaction with PcG, CBX6 modifies chromatin and maintains the transcriptionally repressive state of genes. Members of the chromobox domain protein family are characterized by the presence of a 37 amino acid chromobox (CHRromatin Organization MOdifier) domain. This domain plays a mechanistic role in targeting chromodomain proteins to specific regions of the nucleus.
Specifications
| CBX6 | |
| Polyclonal | |
| Unconjugated | |
| CBX6 | |
| CBX6; chromobox 6; chromobox homolog 6; chromobox protein homolog 6; chromobox-like protein 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23466, 315136, 494448 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95503, Q9DBY5 | |
| CBX6 | |
| A synthesized peptide derived from human CBX6(Accession O95503), corresponding to amino acid residues M1-I48. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction