missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBX7 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530471

Product Code. 13454967

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Human (99%), Rhesus Monkey (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CBX7
Polyclonal
Unconjugated
CBX7
1600014J01Rik; AI851678; Cbx7; chromobox 7; chromobox homolog 7; chromobox protein homolog 7; chromobox-like protein 7; D15Ertd417e
Rabbit
Antigen affinity chromatography
RUO
23492
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.56 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O95931
CBX7
Recombinant protein encompassing a sequence within the center region of human CBX7. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.