missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx8 Antibody (OTI3D1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72087B
This item is not returnable.
View return policy
Description
Cbx8 Monoclonal specifically detects Cbx8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cbx8 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CBX8 | |
| Recombinant protein fragment corresponding to amino acids 1-260 of human Cbx8 (NP_065700) produced in E.coli. | |
| 0.1 mL | |
| 57332.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3D1 | |
| Western Blot, Immunohistochemistry | |
| chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction