missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX8 Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800617S
This item is not returnable.
View return policy
Description
CBX8 Monoclonal antibody specifically detects CBX8 in Canine, Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CBX8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q567U6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 476119, 54520 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D1 | |
| Unconjugated | |
| CCDC93 | |
| CBX8; chromobox 8; chromobox homolog 8; chromobox homolog 8 (Pc class homolog, Drosophila); chromobox protein homolog 8; HPC3; mPc3; Pc class 3 homolog; Pc3; polycomb 3; polycomb 3 homolog; RC1; rectachrome 1 | |
| Human recombinant protein fragment corresponding to amino acids 293-631 of human CCDC93 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction