missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX8 Monoclonal Antibody (OTI3A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800492
This item is not returnable.
View return policy
Description
Chromobox homolog 8 (CBX8 or Pc3) is a member of Drosophila Polycomb group gene family. The polycomb group (PcG) genes are essential for maintece of appropriate expression patterns of developmental master regulators and thus are essential for proper development. Changes in expression of PcG proteins have been associated with cancer. CBX8 is involved in maintaining the transcriptionally repressive state of genes. It modifies chromatin, rendering it heritably changed in its expressibility. Structurally, CBX4 contains 1 chromo domain, which is 40 to 50 amino acids long.Specifications
| CBX8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HC52 | |
| CBX8 | |
| Human recombit protein fragment corresponding to amino acids 1-260 of human CBX8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A11 | |
| Unconjugated | |
| CBX8 | |
| PC3, RC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57332 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction