missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583193
This item is not returnable.
View return policy
Description
Immunogen sequence: LAVDTFPARV IKHRAAFLEA KGQGALDPNG TRVRHGSGPP SSVGGLYRDM GAQGGRPSLI ARIPVARILG DPEEESWSPS LTNLEKVVVT DVTSNFLTVT IKESNTDQGF Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 94%.
Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
Specifications
| CBX8 | |
| Polyclonal | |
| Unconjugated | |
| CBX8 | |
| CBX8; chromobox 8; chromobox homolog 8; chromobox homolog 8 (Pc class homolog, Drosophila); chromobox protein homolog 8; HPC3; mPc3; Pc class 3 homolog; Pc3; polycomb 3; polycomb 3 homolog; RC1; rectachrome 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HC52 | |
| CBX8 | |
| Recombinant protein corresponding to Human CBX8. Recombinant protein control fragment (Product #RP-102525). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction