Learn More
CBY1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12239-1-AP-20UL
This item is not returnable.
View return policy
Description
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| CBY1 | |
| Polyclonal | |
| Unconjugated | |
| CBY1 | |
| arb1, ARPP binding protein, C22orf2, CBY, CBY1, chibby homolog 1 (Drosophila), Cytosolic leucine rich protein, HS508I15A, PGEA1, PIGEA 14, PIGEA14, Protein chibby homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246768, 25776, 73739 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K4I6, Q9D1C2, Q9Y3M2 | |
| Cby1 | |
| CBY1 Fusion Protein Ag2882 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |