Learn More
CBY1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12239-1-AP-20UL
 View more versions of this product
Product Code. 16879833
  • € 145.00 / 20µL
Description
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CBY1
Polyclonal
Unconjugated
CBY1
arb1, ARPP binding protein, C22orf2, CBY, CBY1, chibby homolog 1 (Drosophila), Cytosolic leucine rich protein, HS508I15A, PGEA1, PIGEA 14, PIGEA14, Protein chibby homolog 1
Rabbit
Antigen Affinity Chromatography
RUO
246768, 25776, 73739
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K4I6, Q9D1C2, Q9Y3M2
Cby1
CBY1 Fusion Protein Ag2882
20 μL
Primary
Mouse, Rat, Human