missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CC2D2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560621
This item is not returnable.
View return policy
Description
Immunogen sequence: SVTPNDQCPR AEVSRREDVK KRSVYLKVLF NNKEVSRTVS RPLGADFRVH FGQIFNLQIV NWPESLTLQV YETVGHSSPT LLAEVFLPIP ETTVVTGRAP TEEVEFSSN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants.
Specifications
| CC2D2A | |
| Polyclonal | |
| Unconjugated | |
| CC2D2A | |
| 5730509K17Rik; b2b1035Clo; CC2D2A; coiled-coil and C2 domain containing 2A; coiled-coil and C2 domain-containing protein 2A; JBTS9; KIAA1345; MKS6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57545 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2K1 | |
| CC2D2A | |
| Recombinant protein corresponding to Human CC2D2A. Recombinant protein control fragment (Product #RP-100190). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction