missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCAR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565519
This item is not returnable.
View return policy
Description
Immunogen sequence: KEERDDETEED NNQDEYDPME AEEAEDEEDD RDEEEMTKRD DKRDINRYCK ERPSKDKEKE KTQMITINRD LLMAFVYFDQ SHCGYLLE Highest antigen sequence identity to the following orthologs - mouse 83%, rat 84%.
IFI-44 (interferon-induced protein 44), also known as p44 or MTAP44 (microtubule-associated protein 44), is a 444 amino acid protein that localizes to the cytoplasm and, upon induction by IFN-betas, aggregates to form microtubular structures. Human IFI-44 shares 97% sequence similarity with its chimp counterpart, suggesting a conserved role between species. The gene encoding IFI-44 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
Specifications
| CCAR1 | |
| Polyclonal | |
| Unconjugated | |
| CCAR1 | |
| 2610511G16Rik; 9430036H15Rik; Carp1; CARP-1; Ccar1; Cell cycle and apoptosis regulatory protein 1; cell division cycle and apoptosis regulator 1; cell division cycle and apoptosis regulator protein 1; Death inducer with SAP domain; DIS; MGC44628; OTTHUMP00000019709; RGD1560358; RP11-437A18.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55749 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IX12 | |
| CCAR1 | |
| Recombinant Human CCAR1. Recombinant protein control fragment (Product #RP-106193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction