Learn More
CCAR1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25232-1-AP-20UL
 View more versions of this product
Product Code. 18005275
  • € 145.00 / 20µL
Description
The CCAR1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CCAR1. This antibody recognizes human antigen. The CCAR1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CCAR1
250 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
CARP 1, CARP1, CCAR1, Death inducer with SAP domain, DIS, RP11 437A18.1
20 μL
55749
Store at -20°C. Stable for one year after shipment.