Learn More
CCAR1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25232-1-AP-150UL
This item is not returnable.
View return policy
Description
The CCAR1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CCAR1. This antibody recognizes human antigen. The CCAR1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CCAR1 | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| CARP 1, CARP1, CCAR1, Death inducer with SAP domain, DIS, RP11 437A18.1 | |
| 150 μL | |
| 55749 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |