missing translation for 'onlineSavingsMsg'
Learn More

CCDC109A Antibody (CL3576) - Azide and BSA Free, Novus Biologicals™

Product Code. 30286803 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286803 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30286803 Supplier Novus Biologicals Supplier No. NBP344130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCDC109A Monoclonal antibody specifically detects CCDC109A in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence,KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC109A
Applications Western Blot, Immunocytochemistry/Immunofluorescence, KnockDown
Classification Monoclonal
Clone CL3576
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL
Formulation Lyophilized in 5% Trehalose.
Gene Alias C10orf42, coiled-coil domain containing 109A, coiled-coil domain-containing protein 109A, FLJ46135
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q8NE86, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 90550
Reconstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.