missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC110 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | CCDC110 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CCDC110 Polyclonal antibody specifically detects CCDC110 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| CCDC110 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 256309 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Cancer/testis antigen 52, coiled-coil domain containing 110, coiled-coil domain-containing protein 110, CT52Cancer/testis antigen KM-HN-1, KMHN1KM-HN-1, MGC33607 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCDC110 (NP_689988.1). MSPEKQHREEDEVDSVLLSASKILNSSEGVKESGCSDTEYGCIAESENQIQPQSALKVLQQQLESFQALRMQTLQNVSMVQSEISEILNKSIIEVENPQFSSEKNLVFGTRIEKDLPTENQEENLSMEKSHHFEDSKTLHSVEEKLSGDSVNSLPQSVNVPSQIHSEDTLTLRTSTDNLSSNIIIHPSENSDILKNYNNFYRFLPTAPPNVMSQADTVILDKSKITVPFLKHGFCENLDDICHSIKQMKEELQKSHDGEVALTNELQTLQTDPDVHRNGKYDMSPIHQDKMNFIKEENLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title