Learn More
CCDC124 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27106-1-AP-150UL
This item is not returnable.
View return policy
Description
Required for proper progression of late cytokinetic stages.Specifications
| CCDC124 | |
| Polyclonal | |
| Unconjugated | |
| CCDC124 | |
| CCDC124 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115098, 234388 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96CT7, Q9D8X2 | |
| Ccdc124 | |
| CCDC124 Fusion Protein Ag25930 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |