missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC134 Antibody (04), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06134-50ul
This item is not returnable.
View return policy
Description
CCDC134 Monoclonal antibody specifically detects CCDC134 in Human samples. It is validated for ELISA
Specifications
| CCDC134 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 04 | |
| ELISA 1:1000-1:2000 | |
| coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCDC134 (Uniprot#: Q9H6E4; Met 1-Leu 229). | |
| 50 μg | |
| Cancer | |
| 79879 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction