missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC134 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00083-100ul
This item is not returnable.
View return policy
Description
CCDC134 Polyclonal antibody specifically detects CCDC134 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| CCDC134 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
| coiled-coil domain containing 134, coiled-coil domain-containing protein 134, dJ821D11.3, FLJ22349, MGC21013 | |
| Produced in rabbits immunized with purified, recombinant Human CCDC134 (Uniprot#: Q9H6E4; Met1-Leu229) | |
| 100 μg | |
| Cancer | |
| 79879 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction