missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC134 Monoclonal Antibody (04)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530134
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CCDC134.
The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. One such protein is CCDC134, a recently identified secretory protein that has been found to inhibit the transcriptional activity of the Elk1 protein. Overexpression CCDC134 also inhibited the phosphorylation of Erk and JNK/SAPK but not p38 MAPK, while specific siRNA against CCDC134 activated Elk1 transcriptional activity and the phosphorylation of Erk and JNK/SAPK, suggesting a potential inhibiting role of CCDC134 in MAPK-mediated Elk1 transcription. CCDC134 is widely expressing in normal adult tissues, tumors, and cell lines.
Specifications
| CCDC134 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9H6E4 | |
| CCDC134 | |
| Recombinant Human CCDC134 protein (Met1-Leu229). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 04 | |
| Unconjugated | |
| CCDC134 | |
| 2310042L06Rik; AW208859; Ccdc134; coiled-coil domain containing 134; coiled-coil domain-containing protein 134; dJ821D11.3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction