missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15742
This item is not returnable.
View return policy
Description
CCDC14 Polyclonal specifically detects CCDC14 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CCDC14 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| coiled-coil domain containing 14, DKFZp434L1050, FLJ12892 | |
| Rabbit | |
| 84 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q49A88 | |
| CCDC14 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human CCDC14. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 64770 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction