missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC16 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97135H
This item is not returnable.
View return policy
Description
CCDC16 Polyclonal specifically detects CCDC16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCDC16 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 91603 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| CCDC16, coiled-coil domain containing 16, Coiled-coil domain-containing protein 16, MGC20398, OMCG1, SEL13, sel-13 suppressor of lin-12 homolog, zinc finger protein 830 | |
| Produced in rabbits immunized with E. coli-derived Human CCDC16 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction