missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC18 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78738AF647
This item is not returnable.
View return policy
Description
CCDC18 Polyclonal antibody specifically detects CCDC18 in Human samples. It is validated for Western Blot
Specifications
| CCDC18 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil domain containing 18, coiled-coil domain-containing protein 18, dJ717I23.1, NY-SAR-41, sarcoma antigen NY-SAR-41 | |
| The immunogen for this product maps to a region between residue 225 and 275 of human Coiled-Coil Domain Containing 18 using the numbering given in entry CAI22508.1 (GeneID 343099). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 343099 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction