missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: VDESLFGDIK SPAQGQSDSP IVLLRDKHTL QKTLTALGLD RKPETIQLIT RDMVRELIVP TEDPSGESLI ISPEEFERIK WAS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.
The function remains unknown.
Specifications
Specifications
| Antigen | CCDC19 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CFAP45 |
| Gene Accession No. | Q9UL16 |
| Gene Alias | 1700028D05Rik; CCDC19; CFAP45; cilia and flagella associated protein 45; cilia- and flagella-associated protein 45; coiled-coil domain containing 19; Coiled-coil domain-containing protein 19; coiled-coil domain-containing protein 19, mitochondrial; nasopharyngeal epithelium specific protein 1; nasopharyngeal epithelium specific protein 1 (NESG1); nasopharyngeal epithelium-specific protein 1; NESG1; protein CFAP45, mitochondrial; Unknown (protein for MGC:134208) |
| Gene Symbols | CFAP45 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
