missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC21 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78218AF405
This item is not returnable.
View return policy
Description
CCDC21 Polyclonal antibody specifically detects CCDC21 in Human samples. It is validated for Western Blot
Specifications
| CCDC21 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil domain containing 21, coiled-coil domain-containing protein 21, DKFZp434L0117, DKFZp434P232, FLJ13976, FLJ22000 | |
| The immunogen for this product maps to a region between residue 200 and 250 of human Centrosomal Protein 85kDa using the numbering given in entry NP_073615.2 (GeneID 64793). | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 64793 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction