missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC22 Monoclonal Antibody (OTI2A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811120
This item is not returnable.
View return policy
Description
CCDC22 is a recently identified coiled-coil domain-containing protein that has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. In rat, CCDC22 has been observed to localize in multiple regions of the brain, including the prefrontal and somatosensory cortex, dentate gyrus and thalamus, and in the ipsilateral motor neurons of the spinal cord after sciatic nerve transection suggesting that it may play a role in neuronal injury response. The human CCDC22 gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability (XLID).Specifications
| CCDC22 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60826 | |
| CCDC22 | |
| Full length human recombit protein of human CCDC22 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2A9 | |
| Unconjugated | |
| CCDC22 | |
| CXorf37, JM1, RTSC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 28952 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction