missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21209-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CCDC25 | |
| Polyclonal | |
| Unconjugated | |
| CCDC25 | |
| CCDC25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361059, 55246, 67179 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q78PG9, Q86WR0 | |
| Ccdc25 | |
| CCDC25 Fusion Protein Ag15593 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction