missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC27 Monoclonal Antibody (OTI1F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811190
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| CCDC27 | |
| Monoclonal | |
| Unconjugated | |
| CCDC27 | |
| coiled-coil domain containing 27 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 148870 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1F8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q2M243 | |
| CCDC27 | |
| Full length human recombit protein of human CCDC27 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction