missing translation for 'onlineSavingsMsg'
Learn More

CCDC27 Monoclonal Antibody (OTI4D4), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA811226S

Encompass_Preferred

 View more versions of this product

Product Code. 17555054

  • € 242.00 / 30µL
Only null left
Estimated Shipment: 26-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

CCDC27 Monoclonal antibody specifically detects CCDC27 in Human samples. It is validated for Western Blot
Specifications

Specifications

CCDC27
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BZP6
Mouse
Affinity chromatography
RUO
27159
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI4D4
Unconjugated
CHIA
CCDC27; coiled-coil domain containing 27; coiled-coil domain-containing protein 27; RP1-286D6.1
Full length human recombinant protein of human CHIA produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.