missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (RPA-M1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27215JF646
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CD14 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD14 | |
| Human PHA-stimulated PBMC were used as immunogen. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| RPA-M1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction