missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CCDC28A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17277713 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17277713 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17277713 Supplier Invitrogen™ Supplier No. PA5114952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCDC28A.

This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC28A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CCDC28A
Gene Accession No. Q8IWP9
Gene Alias 1700009P13Rik; AI480677; C6orf80; CCDC28A; CCRL1AP; chemokine C-C motif receptor-like 1 adjacent; coiled-coil domain containing 28A; coiled-coil domain-containing protein 28A; RGD1310326
Gene Symbols CCDC28A
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCDC28A(Accession Q8IWP9), corresponding to amino acid residues N129-E179.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215814, 25901, 361454
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.