missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC28B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531561
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product of this gene localizes to centrosomes and basal bodies. It interacts and colocalizes with several proteins associated with Bardet-Biedl syndrome (BBS).
Specifications
| CCDC28B | |
| Polyclonal | |
| Unconjugated | |
| CCDC28B | |
| 1110002E08Rik; 1810010N17Rik; Ccdc28b; coiled coil domain containing 28B; coiled-coil domain containing 28B; coiled-coil domain-containing protein 28B; RP4-622L5.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BUN5 | |
| CCDC28B | |
| Recombinant protein encompassing a sequence within the center region of human CCDC28B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction