missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC43 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-97844AF488
This item is not returnable.
View return policy
Description
CCDC43 Polyclonal antibody specifically detects CCDC43 in Human samples. It is validated for Western Blot
Specifications
| CCDC43 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil domain containing 43, coiled-coil domain-containing protein 43, FLJ31795 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain-containing protein 43 using the numbering given in entry NP_653210.2 (GeneID 124808). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 124808 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction