missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC53 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24445-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CCDC53 | |
| Polyclonal | |
| Unconjugated | |
| WASHC3 | |
| CCDC53, CGI 116, WASH complex subunit CCDC53, x0009 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51019, 67282 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9CR27, Q9Y3C0 | |
| WASHC3 | |
| CCDC53 Fusion Protein Ag19831 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction