missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598463
This item is not returnable.
View return policy
Description
CCDC57 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CCDC57 (Coiled-Coil Domain Containing 57) is a Protein Coding gene. Diseases associated with CCDC57 include Uterine Benign Neoplasm and Reproductive Organ Benign Neoplasm. An important paralog of this gene is CCHCR1.
Specifications
| CCDC57 | |
| Polyclonal | |
| Unconjugated | |
| CCDC57 | |
| CCDC57; coiled-coil domain containing 57; coiled-coil domain-containing protein 57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284001 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q2TAC2 | |
| CCDC57 | |
| Recombinant Human Coiled-coil domain-containing protein 57 protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction