missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553905
This item is not returnable.
View return policy
Description
Immunogen sequence: FKKIQALQKE KETLAVNYEK EEEFLTNELS RKLMQLQHEK AELEQHLEQE QEFQVNKLMK KIKKLENDTI SKQLTLEQLR REKI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
Specifications
| CCDC6 | |
| Polyclonal | |
| Unconjugated | |
| CCDC6 | |
| 2810012H18Rik; AA536681; AA960498; AW061011; CCDC6; coiled-coil domain containing 6; coiled-coil domain-containing protein 6; D10S170; H4; Papillary thyroid carcinoma-encoded protein; Protein H4; PTC; TPC; TST1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16204 | |
| CCDC6 | |
| Recombinant protein corresponding to Human CCDC6. Recombinant protein control fragment (Product #RP-92555). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction