missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC69 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-77145AF594
This item is not returnable.
View return policy
Description
CCDC69 Polyclonal antibody specifically detects CCDC69 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CCDC69 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 26112 | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| coiled-coil domain containing 69, coiled-coil domain-containing protein 69, DKFZP434C171, FLJ13705 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CCDC69. The immunogen is located within amino acids 150 - 200 of CCDC69. Amino Acid Squence: RNYKKHIQDYGSPSQFWE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction