missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC69 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565748
This item is not returnable.
View return policy
Description
Immunogen sequence: EPHELGPLNGD TAITVQLCAS EEAERHQKDI TRILQQHEEE KKKWAQQVEK ERELELRDRL DEQQRVLEGK NEEALQVLRA Highest antigen sequence identity to the following orthologs - mouse 30%, rat 30%.
May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt].
Specifications
| CCDC69 | |
| Polyclonal | |
| Unconjugated | |
| CCDC69 | |
| 2210021E03Rik; CCDC69; coiled-coil domain containing 69; coiled-coil domain-containing protein 69; D11Ertd461e; RGD1562251 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NI79 | |
| CCDC69 | |
| Recombinant Human CCDC69. Recombinant protein control fragment (Product #RP-104955). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction