missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC7 Polyclonal Antibody

Description
Immunogen sequence: SKSSVKVMLSK TMDKENRPEA VKSCEALAQK IEEFLEAHST DEFKDVSATE PQTAHSMTNR FNAMLKVFEN Q Highest antigen sequence identity to the following orthologs - mouse 40%, rat 42%.
May play a role in tumorigenesis.
Specifications
Specifications
| Antigen | CCDC7 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCDC7 |
| Gene Accession No. | Q96M83 |
| Gene Alias | BIOT2; BioT2-A; BioT2-B; BioT2-C; C10orf68; CCDC7; coiled-coil domain containing 7; Coiled-coil domain-containing protein 7; coiled-coil domain-containing protein 7; uncharacterized protein C10orf68; Protein BIOT2; RP11-479G22.1 |
| Gene Symbols | CCDC7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?