missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC79 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144158
This item is not returnable.
View return policy
Description
CCDC79 Polyclonal Antibody for IHC (P), ELISA
Meiosis-specific telomere-associated protein involved in meiotic telomere attachment to the nucleus inner membrane, a crucial step for homologous pairing and synapsis. Component of the MAJIN-TERB1-TERB2 complex, which promotes telomere cap exchange by mediating attachment of telomeric DNA to the inner nuclear membrane and replacement of the protective cap of telomeric chromosomes: in early meiosis, the MAJIN-TERB1-TERB2 complex associates with telomeric DNA and the shelterin/telosome complex. During prophase, the complex matures and promotes release of the shelterin/telosome complex from telomeric DNA. In the MAJIN-TERB1-TERB2 complex, TERB1 probably mediates association with the shelterin/telosome complex via interaction with TERF1, promoting priming telomeric DNA attachment'. Promotes telomere association with the nuclear envelope and deposition of the SUN-KASH/LINC complex. Also recruits cohesin to telomeres to develop structural rigidity.
Specifications
| CCDC79 | |
| Polyclonal | |
| Unconjugated | |
| TERB1 | |
| CCDC79; coiled-coil domain containing 79; coiled-coil domain-containing protein 79; telomere repeat binding bouquet formation protein 1; telomere repeats-binding bouquet formation protein 1; TERB1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 283847 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q8NA31 | |
| TERB1 | |
| Recombinant Human Telomere repeats-binding bouquet formation protein 1 protein (301-550AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction