Learn More
CCDC90B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-93528
Product Code. 18737283
  • € 572.00 / 0.10mL
Description
CCDC90B Polyclonal specifically detects CCDC90B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry
coiled-coil domain containing 90B, mitochondrial
Human, Rat
IgG
Polyclonal
Affinity Purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.